Ethyl decanoate ≥98.0%

Supplier: TCI America

D0022-500ML D0022-25ML
TCD0022-500MLEA 123.24 CAD
TCD0022-500ML TCD0022-025ML
Ethyl decanoate ≥98.0%
Ethyl decanoate

CAS Number: 110-38-3
MDL Number: MFCD00009581
Molecular Formula: C12H24O2
Molecular Weight: 200.32
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 243
Melting point (°C): -20
Flash Point (°C): 123
Specific Gravity (20/20): 0.86


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Decanoic Acid Ethyl Ester

Formula: C₁₂H₂₄O₂
Boiling Pt: 243 °C
Melting Pt: –20 °C
Density: 0.86 (20 °C)
Flash Pt: 123 °C
MDL Number: MFCD00009581
CAS Number: 110-38-3
EINECS: 203-761-9
Merck Index: 13,03811
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e07a4c3d5abc6810e3bd9e8b7c86e8bf2b3d3f&vid=c4e07a4c3d5abc6810e3bd9e8b7c86e8bf2b3d3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR