Capric acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
D0017-25G
D0017-400G
TCD0017-025GEA
56.53
CAD
TCD0017-025G
TCD0017-400G
Capric acid ≥98.0% (by GC, titration analysis)
Capric acid
CAS Number: 334-48-5
MDL Number: MFCD00004441
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 270
Flash Point (°C): 150
Freezing point (°C): 30
Formula:
C₁₀H₂₀O₂ Boiling Pt: 270 °C Melting Pt: 29…32 °C Density: 0.893 g/cm³ (20 °C) Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004441 CAS Number: 334-48-5 EINECS: 206-376-4 Merck Index: 13,01764 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...