1,10-Decanediol ≥97.0%

Supplier: TCI America

Synonyms: 1,10-Dihydroxy decane

D0014-25G D0014-500G D0014-100G
TCD0014-025GEA 31.81 CAD
TCD0014-025G TCD0014-500G TCD0014-100G
1,10-Decanediol ≥97.0%
1,10-Decanediol

CAS Number: 112-47-0
MDL Number: MFCD00004749
Molecular Formula: C10H22O2
Molecular Weight: 174.28
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 192
Melting point (°C): 73
Flash Point (°C): 160


Other Product Names / Synonyms: Decamethylene Glycol, 1,10-Dihydroxydecane

Formula: C₁₀H₂₂O₂
MW: 174.28 g/mol
Boiling Pt: 192 °C
Melting Pt: 73 °C
Density: 0.883 g/cm³ (80 °C)
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004749
CAS Number: 112-47-0
EINECS: 203-975-2
Merck Index: 13,02869
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3ba47b5ac061eeecaa6a3795b80f1c3a12c9bc3&vid=c7029b91d2fe6e0d125c6cc73e2f3619bf8acb99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR