1,12-Dodecanedioic acid ≥99.0%

Supplier: TCI America
Warning

Synonyms: Dodecanedioic acid, 1,10-Decanedicarboxylic acid

D0013-25G D0013-500G
TCD0013-025GEA 41.33 CAD
TCD0013-025G TCD0013-500G
1,12-Dodecanedioic acid ≥99.0%
1,12-Dodecanedioic acid

CAS Number: 693-23-2
MDL Number: MFCD00002735
Molecular Formula: C12H22O4
Molecular Weight: 230.30
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 129
Flash Point (°C): 227


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,10-Decanedicarboxylic Acid

Formula: C₁₂H₂₂O₄
Boiling Pt: 294 °C (1013 hPa)
Melting Pt: 129 °C
Density: 1.175 g/cm³ (21 °C)
Flash Pt: 227 °C
Storage Temperature: Freezer
MDL Number: MFCD00002735
CAS Number: 693-23-2
EINECS: 211-746-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8628028465cd4580e57b78121afe3b1053edf93f&vid=8628028465cd4580e57b78121afe3b1053edf93f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR