5,5-Dimethyloxazolidine-2,4-dione (dimethadione) ≥99.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Dimethadione, 5,5-Dimethyloxazolidine-2,4-dione
D0003-25G
D0003-5G
TCD0003-25GEA
575.28
CAD
TCD0003-25G
TCD0003-005G
5,5-Dimethyloxazolidine-2,4-dione (dimethadione) ≥99.0% (by GC, titration analysis)
5,5-Dimethyloxazolidine-2,4-dione (Dimethadione)
CAS Number: 695-53-4
MDL Number: MFCD00005379
Molecular Formula: C5H7NO3
Molecular Weight: 129.12
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 137
Melting point (°C): 79
Other Product Names / Synonyms: Dimethadione
Formula:
C₅H₇NO₃ MW: 129.12 g/mol Boiling Pt: 137 °C Melting Pt: 79 °C |
MDL Number:
MFCD00005379 CAS Number: 695-53-4 EINECS: 211-781-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...