2-Chloroethyl toluene-4-sulfonate ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloroethyl p-toluenesulphonate, 2-Chloroethyl p-toluenesulfonate

C2380-100G C2380-25G
TCC2380-100GEA 233.84 CAD
TCC2380-100G TCC2380-25G
2-Chloroethyl toluene-4-sulfonate ≥98.0%
2-Chloroethyl toluene-4-sulphonate

CAS Number: 80-41-1
MDL Number: MFCD00000970
Molecular Formula: C9H11ClO3S
Molecular Weight: 234.69
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 156
Flash Point (°C): 113
Freezing point (°C): 20
Specific Gravity (20/20): 1.30


Other Product Names / Synonyms: p-Toluenesulfonic Acid 2-Chloroethyl Ester

Formula: C₉H₁₁ClO₃S
MW: 234.7 g/mol
Boiling Pt: 156 °C
Density: 1.30 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00000970
CAS Number: 80-41-1
EINECS: 201-277-2
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c49127d23420b71a5f2f4654951b607b6e4d0b41&vid=c49127d23420b71a5f2f4654951b607b6e4d0b41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR