Cesium carbonate 98,0

Supplier: TCI America
Warning

C2160-100G C2160-25G
TCC2160-100GEA 346.73 CAD
TCC2160-100G TCC2160-25G
Cesium carbonate 98,0
Cesium carbonate

CAS Number: 534-17-8
MDL Number: MFCD00010957
Molecular Formula: CCs2O3
Molecular Weight: 325.82
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 610

Formula: Cs₂CO₃
MW: 325.82 g/mol
Melting Pt: 610
Density: 4.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9
Merck Index: 13,02020
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7472316f81f62f08637fdd3240f3838174194a5d&vid=7472316f81f62f08637fdd3240f3838174194a5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR