3'-Chloropropiophenone ≥98.0%

Supplier: TCI America

C2033-250G C2033-25G
TCC2033-250GEA 273.34 CAD
TCC2033-250G TCC2033-25G
3'-Chloropropiophenone ≥98.0%
3'-Chloropropiophenone

CAS Number: 34841-35-5
Molecular Formula: C9H9ClO
Molecular Weight: 168.62
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 124
Melting point (°C): 47
Flash Point (°C): 113


Other Product Names / Synonyms: 3-Chlorophenyl Ethyl Ketone

Formula: C₉H₉ClO
Boiling Pt: 124 °C
Melting Pt: 47 °C
Flash Pt: 113 °C
MDL Number: MFCD00009925
CAS Number: 34841-35-5
EINECS: 252-242-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46a5090045db0c50a683865980f050238a3ee949&vid=46a5090045db0c50a683865980f050238a3ee949&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR