N-Cyclohexyl-p-toluenesulfonamide ≥99.0% (by HPLC, total nitrogen)
Supplier: TCI America
C2032-25G
C2032-500G
TCC2032-25GEA
47.4
CAD
TCC2032-25G
TCC2032-500G
N-Cyclohexyl-p-toluenesulfonamide ≥99.0% (by HPLC, total nitrogen)
N-Cyclohexyl-p-toluenesulphonamide
CAS Number: 80-30-8
Molecular Formula: C13H19NO2S
Molecular Weight: 253.36
Purity/Analysis Method: >99.0% (HPLC,N)
Form: Crystal
Boiling point (°C): 350
Melting point (°C): 87
Formula:
C₁₃H₁₉NO₂S MW: 253.3605 g/mol Boiling Pt: 350 °C Melting Pt: 87 °C |
MDL Number:
MFCD00014285 CAS Number: 80-30-8 EINECS: 201-268-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...