2-Chlorobenzenesulfonamide ≥98.0%

Supplier: TCI America

C1990-25G C1990-500G
TCC1990-25GEA 109.02 CAD
TCC1990-25G TCC1990-500G
2-Chlorobenzenesulfonamide ≥98.0%
2-Chlorobenzenesulphonamide

CAS Number: 6961-82-6
Molecular Formula: C6H6ClNO2S
Molecular Weight: 191.63
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 190

Formula: C₆H₆ClNO₂S
Melting Pt: 190 °C
MDL Number: MFCD00051974
CAS Number: 6961-82-6
EINECS: 230-156-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc9319072e0d4d5f93cc88f07c9c4c20b997eec&vid=5fc9319072e0d4d5f93cc88f07c9c4c20b997eec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR