3-Chlorobenzophenone ≥98.0%

Supplier: TCI America

C1775-25G C1775-500G
TCC1775-25GEA 52.14 CAD
TCC1775-25G TCC1775-500G
3-Chlorobenzophenone ≥98.0%
3-Chlorobenzophenone

CAS Number: 1016-78-0
MDL Number: MFCD00009816
Molecular Formula: C13H9ClO
Molecular Weight: 216.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 332
Melting point (°C): 83

Formula: C₁₃H₉ClO
Boiling Pt: 332 °C
Melting Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00009816
CAS Number: 1016-78-0
EINECS: 213-809-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc65adcca8938560f1316e0d361039fec902e326&vid=bc65adcca8938560f1316e0d361039fec902e326&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR