2-Chloro-4,6-dimethoxy-1,3,5-triazine ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

C1676-5G C1676-250G C1676-25G
TCC1676-5GEA 33.18 CAD
TCC1676-5G TCC1676-250G TCC1676-25G
2-Chloro-4,6-dimethoxy-1,3,5-triazine ≥97.0% (by HPLC, titration analysis)
2-Chloro-4,6-dimethoxy-1,3,5-triazine

CAS Number: 3140-73-6
MDL Number: MFCD00075607
Molecular Formula: C5H6ClN3O2
Molecular Weight: 175.57
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 78


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis
Other Product Names / Synonyms: CDMT

Formula: C₅H₆ClN₃O₂
Boiling Pt: 100 °C
Melting Pt: 78 °C
MDL Number: MFCD00075607
CAS Number: 3140-73-6
EINECS: 221-541-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b4584948db56d36ac5a3750515069ef279c934a&vid=0b4584948db56d36ac5a3750515069ef279c934a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR