4-Chloro-2-fluorobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C1668-5G C1668-25G C1668-1G
TCC1668-005GEA 67.94 CAD
TCC1668-005G TCC1668-25G TCC1668-001G
4-Chloro-2-fluorobenzoic acid ≥98.0% (by titrimetric analysis)
4-Chloro-2-fluorobenzoic acid

CAS Number: 446-30-0
MDL Number: MFCD00042468
Molecular Formula: C7H4ClFO2
Molecular Weight: 174.56
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 206

Formula: C₇H₄ClFO₂
Melting Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00042468
CAS Number: 446-30-0
EINECS: 207-162-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96120d3dfba6b1a99a09a0d4af22565006bd4f13&vid=2c3f9b40d4ae492ae3f7125721531fc2937afaab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR