(R)-(-)-2-Chloromandelic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C1663-5G C1663-1G
TCC1663-005GEA 94.8 CAD
TCC1663-005G TCC1663-001G
(R)-(-)-2-Chloromandelic acid ≥98.0% (by titrimetric analysis)
(R)-(-)-2-Chloromandelic acid

CAS Number: 52950-18-2
MDL Number: MFCD00798429
Molecular Formula: C8H7ClO3
Molecular Weight: 186.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 120
Specific rotation [a]20/D: -124.5 deg (C=3, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-2'-Chloro-alpha-hydroxyphenylacetic Acid

Formula: C₈H₇ClO₃
Melting Pt: 120 °C
MDL Number: MFCD00798429
CAS Number: 52950-18-2
EINECS: 444-040-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6181db597d7d0dace190df4c387371a0bc828a93&vid=6181db597d7d0dace190df4c387371a0bc828a93&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR