(1S)-(+)-Camphorquinone ≥97.0%

Supplier: TCI America

Synonyms: (1S)-(+)-Camphorquin

C1660-5G C1660-1G
TCC1660-005GEA 274.92 CAD
TCC1660-005G TCC1660-1G
(1S)-(+)-Camphorquinone ≥97.0%
(1S)-(+)-Camphorquinone

CAS Number: 2767-84-2
MDL Number: MFCD00064159
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 200
Specific rotation [a]20/D: 100 deg (C=1, Toluene)


Product Categories: Bioscience, Bicyclic Monoterpenes
Other Product Names / Synonyms: (1S)-(+)-2,3-Bornanedione

Formula: C₁₀H₁₄O₂
Melting Pt: 200 °C
MDL Number: MFCD00064159
CAS Number: 2767-84-2
EINECS: 220-446-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ee37e1c176d41e68d40da0eb3b12b341695a61f&vid=2ee37e1c176d41e68d40da0eb3b12b341695a61f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR