2-Chloro-6-methylpyridine ≥98.0%

Supplier: TCI America
Warning

C1654-25G C1654-5G
TCC1654-025GEA 150.1 CAD
TCC1654-025G TCC1654-005G
2-Chloro-6-methylpyridine ≥98.0%
2-Chloro-6-methylpyridine

CAS Number: 18368-63-3
MDL Number: MFCD00006245
Molecular Formula: C6H6ClN
Molecular Weight: 127.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 73
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: 6-Chloro-2-picoline

Formula: C₆H₆ClN
Boiling Pt: 88 °C
Density: 1.15 (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00006245
CAS Number: 18368-63-3
EINECS: 242-241-6
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=252897c34aa868ceee48b4b0bc9681d3031dd3a3&vid=252897c34aa868ceee48b4b0bc9681d3031dd3a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR