5-Chloro-1-indanone ≥97.0%
Supplier: TCI America
C1644-1G
C1644-5G
TCC1644-001GEA
104.28
CAD
TCC1644-001G
TCC1644-005G
5-Chloro-1-indanone ≥97.0%
5-Chloro-1-indanone
CAS Number: 42348-86-7
MDL Number: MFCD00041456
Molecular Formula: C9H7ClO
Molecular Weight: 166.60
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 94
Formula:
C₉H₇ClO MW: 166.61 g/mol Boiling Pt: 150 °C (0.2 mmHg) Melting Pt: 94 °C |
MDL Number:
MFCD00041456 CAS Number: 42348-86-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...