3-Chlorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 3-Chlorobenzeneboronic acid, (3-Chlorophenyl)metaboric acid, 3-Chlorophenyl boronic acid, Dihydroxy(3-chlorophenyl)borane

C1613-25G C1613-5G
TCC1613-25GEA 225.94 CAD
TCC1613-25G TCC1613-005G
3-Chlorophenylboronic acid (contains varying amounts of Anhydride)
3-Chlorophenylboronic acid

CAS Number: 63503-60-6
MDL Number: MFCD00161354
Molecular Formula: C6H6BClO2
Molecular Weight: 156.37
Form: Crystal
Color: White


Other Product Names / Synonyms: 3-Chlorobenzeneboronic Acid

Formula: C₆H₆BClO₂
Storage Temperature: Ambient
MDL Number: MFCD00161354
CAS Number: 63503-60-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b055b20cffb6195b7dfeb1646332cb030023025f&vid=b055b20cffb6195b7dfeb1646332cb030023025f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR