4-Chloro-2-fluorobenzaldehyde ≥98.0%

Supplier: TCI America
Warning

C1597-5G C1597-25G
TCC1597-005GEA 127.98 CAD
TCC1597-005G TCC1597-25G
4-Chloro-2-fluorobenzaldehyde ≥98.0%
4-Chloro-2-fluorobenzaldehyde

CAS Number: 61072-56-8
MDL Number: MFCD00143282
Molecular Formula: C7H4ClFO
Molecular Weight: 158.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 60
Storage Temperature: 0-10°C

Formula: C₇H₄ClFO
MW: 158.56 g/mol
Melting Pt: 60 °C
MDL Number: MFCD00143282
CAS Number: 61072-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba071de1f3c9fca27d2cb539aff8b9a215f452ae&vid=a935f654c9dbca28350cee0791bbcc3b232eabd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR