5-Chloro-2-fluoroaniline ≥97.0%
Supplier: TCI America
|
Warning
|
C1596-5G
C1596-25G
TCC1596-005GEA
67.78
CAD
TCC1596-005G
TCC1596-025G
5-Chloro-2-fluoroaniline ≥97.0%
5-Chloro-2-fluoroaniline
CAS Number: 2106-05-0
MDL Number: MFCD00069416
Molecular Formula: C6H5ClFN
Molecular Weight: 145.56
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 103
Specific Gravity (20/20): 1.33
Formula:
C₆H₅ClFN Boiling Pt: 105 °C Density: 1.33 (20 °C) Flash Pt: 103 °C |
MDL Number:
MFCD00069416 CAS Number: 2106-05-0 EINECS: 218-284-1 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...