3-Chloro-2-methylanisole ≥96.0%

Supplier: TCI America

C1585-1G C1585-5G
TCC1585-001GEA 41.08 CAD
TCC1585-001G TCC1585-005G
3-Chloro-2-methylanisole ≥96.0%
3-Chloro-2-methylanisole

CAS Number: 3260-88-6
MDL Number: MFCD00070772
Molecular Formula: C8H9ClO
Molecular Weight: 156.61
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 207
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: 3-Chloro-2-methylanisole

Formula: C₈H₉ClO
Boiling Pt: 207 °C
Density: 1.16 (20 °C)
MDL Number: MFCD00070772
CAS Number: 3260-88-6
EINECS: 221-862-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cf81bb34ef29d4b2cc7a4f5c1b183e2ada7ef04&vid=d5a553510d0f548de0fb55f434db449fe08dc62a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR