3-Hydroxybenzonitrile ≥98.0%

Supplier: TCI America

C1583-25G C1583-5G
TCC1583-25GEA 235.42 CAD
TCC1583-25G TCC1583-005G
3-Hydroxybenzonitrile ≥98.0%
3-Hydroxybenzonitrile

CAS Number: 873-62-1
MDL Number: MFCD00002252
Molecular Formula: C7H5NO
Molecular Weight: 119.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 80


Other Product Names / Synonyms: 3-Hydroxybenzonitrile

Formula: C₇H₅NO
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00002252
CAS Number: 873-62-1
EINECS: 212-845-4
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20d6c4cd264da1a0718db13e2c1f63c6372fa80c&vid=49d1bfa8bd92fab172665764989594539be25c30&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR