(4-Chlorobenzyl)triphenylphosphonium chloride ≥98.0%
Supplier: TCI America
C1581-5G
C1581-25G
TCC1581-005GEA
41.08
CAD
TCC1581-005G
TCC1581-025G
(4-Chlorobenzyl)triphenylphosphonium chloride ≥98.0%
(4-Chlorobenzyl)triphenylphosphonium chloride
CAS Number: 1530-39-8
MDL Number: MFCD00041533
Molecular Formula: C25H21Cl2P
Molecular Weight: 423.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Formula:
C₂₅H₂₁Cl₂P |
MDL Number:
MFCD00041533 CAS Number: 1530-39-8 EINECS: 216-228-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...