1-Chloro-2,5-dimethoxybenzene ≥97.0%

Supplier: TCI America

Synonyms: 2-Chloro-1,4-dimethoxybenzene

C1577-500G C1577-25G
TCC1577-500GEA 276.03 CAD
TCC1577-500G TCC1577-025G
1-Chloro-2,5-dimethoxybenzene ≥97.0%
1-Chloro-2,5-dimethoxybenzene

CAS Number: 2100-42-7
MDL Number: MFCD00008356
Molecular Formula: C8H9ClO2
Molecular Weight: 172.61
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 123
Specific Gravity (20/20): 1.23


Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 2-Chloro-1,4-dimethoxybenzene

Formula: C₈H₉ClO₂
MW: 172.61 g/mol
Boiling Pt: 235 °C
Melting Pt: 8.1 °C
Density: 1.23 (20 °C)
Flash Pt: 123 °C
MDL Number: MFCD00008356
CAS Number: 2100-42-7
EINECS: 218-267-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4793e7952f0ed857061dc6007dbf5967f52dc987&vid=4793e7952f0ed857061dc6007dbf5967f52dc987&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR