β-Chloro-4'-fluoropropiophenone ≥96.0%

Supplier: TCI America

C1570-25G C1570-5G
TCC1570-25GEA 184.86 CAD
TCC1570-25G TCC1570-005G
β-Chloro-4'-fluoropropiophenone ≥96.0%
β-Chloro-4'-fluoropropiophenone

CAS Number: 347-93-3
MDL Number: MFCD00000991
Molecular Formula: C9H8ClFO
Molecular Weight: 186.61
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 103
Melting point (°C): 47
Flash Point (°C): 92

Formula: C₉H₈ClFO
Boiling Pt: 103 °C
Melting Pt: 47 °C
Flash Pt: 92 °C
MDL Number: MFCD00000991
CAS Number: 347-93-3
EINECS: 206-475-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a112fc50c34c76d7610b495ca8b3470ae17c403&vid=6a112fc50c34c76d7610b495ca8b3470ae17c403&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR