Benzyl carbazate ≥98.0%

Supplier: TCI America

Synonyms: Hydrazine, N-CBZ protected, Benzyl hydrazinecarboxylate

C1564-25G C1564-5G
TCC1564-25GEA 79 CAD
TCC1564-25G TCC1564-005G
Benzyl carbazate ≥98.0%
Benzyl carbazate

CAS Number: 5331-43-1
MDL Number: MFCD00041890
Molecular Formula: C8H10N2O2
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 69
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis
Other Product Names / Synonyms: Carbazic Acid Benzyl Ester, Carbobenzoxyhydrazine, N-Cbz-hydrazine

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 69 °C
Flash Pt: 110 °C
MDL Number: MFCD00041890
CAS Number: 5331-43-1
EINECS: 226-230-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f80f7f1f4dcda73073fd17ca913bc6c0a66dbe2&vid=6f80f7f1f4dcda73073fd17ca913bc6c0a66dbe2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR