6-Chloro-m-anisidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C1555-25G C1555-5G
TCC1555-25GEA 2711.28 CAD
TCC1555-25G TCC1555-005G
6-Chloro-m-anisidine hydrochloride ≥98.0% (by HPLC, titration analysis)
6-Chloro-m-anisidine hydrochloride

CAS Number: 85006-21-9
MDL Number: MFCD00012962
Molecular Formula: C7H8ClNO
Molecular Weight: 194.06
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 6-Chloro-m-anisidine Hydrochloride, 3-Amino-4-chloroanisole Hydrochloride, 4-Chloro-3-aminoanisole Hydrochloride

Formula: C₇H₉Cl₂NO
Melting Pt: 208 °C
MDL Number: MFCD00012962
CAS Number: 85006-21-9
EINECS: 285-050-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd09107410892f1359208c34425303d990c2e5ef&vid=dd09107410892f1359208c34425303d990c2e5ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR