4-Chloro-3-nitrobenzonitrile ≥98.0%

Supplier: TCI America

C1548-25G C1548-5G
TCC1548-025GEA 94.8 CAD
TCC1548-025G TCC1548-005G
4-Chloro-3-nitrobenzonitrile ≥98.0%
4-Chloro-3-nitrobenzonitrile

CAS Number: 939-80-0
MDL Number: MFCD00016987
Molecular Formula: C7H3ClN2O2
Molecular Weight: 182.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 99

Formula: C₇H₃ClN₂O₂
Melting Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00016987
CAS Number: 939-80-0
EINECS: 213-364-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a58331d7aab78cbd86f674f89732482ac3609d5&vid=5af5e8438d7d2e8bcaa986c6dfc37cf23e4fbdcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR