4-Chloro-3-(trifluoromethyl)phenol ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloro-5-hydroxybenzotrifluoride

C1516-1G C1516-5G
TCC1516-001GEA 93.22 CAD
TCC1516-001G TCC1516-005G
4-Chloro-3-(trifluoromethyl)phenol ≥98.0%
4-Chloro-3-(trifluoromethyl)phenol

CAS Number: 6294-93-5
MDL Number: MFCD00042525
Molecular Formula: C7H4ClF3O
Molecular Weight: 196.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 122
Melting point (°C): 43
Flash Point (°C): 83


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Chloro-3-(trifluoromethyl)phenol

Formula: C₇H₄ClF₃O
Boiling Pt: 122 °C
Melting Pt: 43 °C
Flash Pt: 83 °C
MDL Number: MFCD00042525
CAS Number: 6294-93-5
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bab85afd0ab0600854093d7827f1f22524c05010&vid=bab85afd0ab0600854093d7827f1f22524c05010&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR