3-Chloro-4-fluorobenzotrifluoride ≥98.0%

Supplier: TCI America

C1515-25G C1515-5G
TCC1515-25GEA 92.59 CAD
TCC1515-25G TCC1515-005G
3-Chloro-4-fluorobenzotrifluoride ≥98.0%
3-Chloro-4-fluorobenzotrifluoride

CAS Number: 78068-85-6
MDL Number: MFCD00042207
Molecular Formula: C7H3ClF4
Molecular Weight: 198.54
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.45
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Chloro-alpha,alpha,alpha,4-tetrafluorotoluene

Formula: C₇H₃ClF₄
Density: 1.45 (20 °C)
MDL Number: MFCD00042207
CAS Number: 78068-85-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e61bfa960a93aca75bfa913bd49d406834dfe07&vid=8e61bfa960a93aca75bfa913bd49d406834dfe07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR