5-Chlorooxindole ≥97.0% (by GC, total nitrogen)

Supplier: TCI America

Synonyms: 5-Chloro-2-oxindole

C1509-1G C1509-5G
TCC1509-1GEA 30.02 CAD
TCC1509-1G TCC1509-005G
5-Chlorooxindole ≥97.0% (by GC, total nitrogen)
5-Chlorooxindole

CAS Number: 17630-75-0
MDL Number: MFCD00191927
Molecular Formula: C8H6ClNO
Molecular Weight: 167.59
Purity/Analysis Method: >97.0% (GC,N)
Form: Crystal
Melting point (°C): 197


Other Product Names / Synonyms: 5-Chloro-2-indolinone

Formula: C₈H₆ClNO
MW: 167.59 g/mol
Melting Pt: 197 °C
MDL Number: MFCD00191927
CAS Number: 17630-75-0
EINECS: 412-200-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d963fc3a71bc7a7af970141bdbf74b2047305953&vid=d963fc3a71bc7a7af970141bdbf74b2047305953&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR