4-Chloro-3-fluorobenzaldehyde ≥97.0%

Supplier: TCI America
Warning

C1506-1G C1506-25G C1506-5G
TCC1506-1GEA 66.36 CAD
TCC1506-1G TCC1506-25G TCC1506-005G
4-Chloro-3-fluorobenzaldehyde ≥97.0%
4-Chloro-3-fluorobenzaldehyde

CAS Number: 5527-95-7
MDL Number: MFCD00143288
Molecular Formula: C7H4ClFO
Molecular Weight: 158.56
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 48
Storage Temperature: 0-10°C

Formula: C₇H₄ClFO
Melting Pt: 48 °C
MDL Number: MFCD00143288
CAS Number: 5527-95-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ecc1102a007e11c696612722143de63ce081fe8&vid=a769f30533d7612c27ef32d2c32365de14256ec4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR