1-Chloro-3-fluoro-4-iodobenzene ≥98.0%

Supplier: TCI America

Synonyms: 5-Chloro-1-fluoro-2-iodobenzene, 4-Chloro-2-fluoro-1-iodobenzene, 4-Chloro-2-fluoroiodobenzene

C1499-25G C1499-5G
TCC1499-25GEA 281.24 CAD
TCC1499-25G TCC1499-005G
1-Chloro-3-fluoro-4-iodobenzene ≥98.0%
1-Chloro-3-fluoro-4-iodobenzene

CAS Number: 6797-79-1
MDL Number: MFCD00079729
Molecular Formula: C6H3ClFI
Molecular Weight: 256.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Freezing point (°C): 18
Specific Gravity (20/20): 2.01

Formula: C₆H₃ClFI
Density: 2.01 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00079729
CAS Number: 6797-79-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bc91475b8bd4412742d63f0dd62426055507d59&vid=0bc91475b8bd4412742d63f0dd62426055507d59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR