3-Chloro-4-fluorobenzaldehyde ≥98.0%

Supplier: TCI America
Warning

C1498-5G C1498-25G
TCC1498-005GEA 33.18 CAD
TCC1498-005G TCC1498-25G
3-Chloro-4-fluorobenzaldehyde ≥98.0%
3-Chloro-4-fluorobenzaldehyde

CAS Number: 34328-61-5
MDL Number: MFCD00011735
Molecular Formula: C7H4ClFO
Molecular Weight: 158.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 24

Formula: C₇H₄ClFO
Boiling Pt: 66 °C (2 mmHg)
Melting Pt: 44…46 °C
Flash Pt: 110 °C
MDL Number: MFCD00011735
CAS Number: 34328-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0fe1cb982861f61c3abf6c1c98f533d16ea488b&vid=ca44732308e7fd34bae4516d39a7cc3231a897e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR