Indole-5-carbonitrile ≥98.0%
Supplier: TCI America
Synonyms:
1H-Indole-5-carbonitrile, 5-Cyanoindole, 5-Cyano-1H-indole
C1491-1G
C1491-5G
C1491-25G
TCC1491-001GEA
41.08
CAD
TCC1491-001G
TCC1491-005G
TCC1491-25G
Indole-5-carbonitrile ≥98.0%
Indole-5-carbonitrile
CAS Number: 15861-24-2
MDL Number: MFCD00005669
Molecular Formula: C9H6N2
Molecular Weight: 142.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 106
Other Product Names / Synonyms: 5-Indolecarbonitrile
Formula:
C₉H₆N₂ Melting Pt: 106 °C |
MDL Number:
MFCD00005669 CAS Number: 15861-24-2 EINECS: 239-986-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...