2-Chloro-4-methoxyphenol ≥98.0%
Supplier: TCI America
Synonyms:
3-Chlor-4-hydroxyanisole
C1487-5G
C1487-25G
TCC1487-005GEA
244.9
CAD
TCC1487-005G
TCC1487-025G
2-Chloro-4-methoxyphenol ≥98.0%
2-Chloro-4-methoxyphenol
CAS Number: 18113-03-6
MDL Number: MFCD00070773
Molecular Formula: C7H7ClO2
Molecular Weight: 158.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 43
Flash Point (°C): 85
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 3-Chloro-4-hydroxyanisole
Formula:
C₇H₇ClO₂ MW: 158.58 g/mol Boiling Pt: 242 °C (1013 hPa) Melting Pt: 43 °C Flash Pt: 85 °C |
MDL Number:
MFCD00070773 CAS Number: 18113-03-6 EINECS: 242-007-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...