(1R)-(-)-Camphorquinone ≥98.0%

Supplier: TCI America

Synonyms: (1R,4S)-1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione

C1482-5G C1482-1G
TCC1482-005GEA 334.96 CAD
TCC1482-005G TCC1482-1G
(1R)-(-)-Camphorquinone ≥98.0%
(1R)-(-)-Camphorquinone

CAS Number: 10334-26-6
MDL Number: MFCD00082863
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 200
Specific rotation [a]20/D: -100 deg (C=1, Toluene)


Product Categories: Bioscience, Bicyclic Monoterpenes
Other Product Names / Synonyms: (1R)-(-)-2,3-Bornanedione

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00082863
CAS Number: 10334-26-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e89d10c89e5d6ce19f47fe59b7c5c0693c565692&vid=e89d10c89e5d6ce19f47fe59b7c5c0693c565692&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR