4-Chlorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Chlorobenzeneboronic acid, Dihydroxy(4-chlorophenyl)borane, (4-Chlorophenyl)boronic acid, (4-Chlorophenyl)metaboric acid
C1473-5G
C1473-25G
TCC1473-005GEA
78.78
CAD
TCC1473-005G
TCC1473-25G
4-Chlorophenylboronic acid (contains varying amounts of Anhydride)
4-Chlorophenylboronic acid
CAS Number: 1679-18-1
MDL Number: MFCD00039137
Molecular Formula: C6H6BClO2
Molecular Weight: 156.37
Form: Crystal
Color: White
Other Product Names / Synonyms: 4-Chlorobenzeneboronic Acid
Formula:
C₆H₆BClO₂ Melting Pt: 268…272 °C Density: ∼1 g/cm³ (20 °C) |
MDL Number:
MFCD00039137 CAS Number: 1679-18-1 EINECS: 216-845-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...