2-Chloro-4-fluorobenzaldehyde ≥97.0%

Supplier: TCI America
Warning

C1465-5G C1465-1G
TCC1465-005GEA 53.04 CAD
TCC1465-005G TCC1465-001G
2-Chloro-4-fluorobenzaldehyde ≥97.0%
2-Chloro-4-fluorobenzaldehyde

CAS Number: 84194-36-5
MDL Number: MFCD00042527
Molecular Formula: C7H4ClFO
Molecular Weight: 158.56
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 61
Flash Point (°C): 110

Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 61 °C
Flash Pt: 110 °C
MDL Number: MFCD00042527
CAS Number: 84194-36-5

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR