2-Chloro-4,5-difluorobenzoic acid ≥98.0%

Supplier: TCI America
Danger

C1460-25G C1460-250G
TCC1460-025GEA 70.74 CAD
TCC1460-025G TCC1460-250G
2-Chloro-4,5-difluorobenzoic acid ≥98.0%
2-Chloro-4,5-difluorobenzoic acid

CAS Number: 110877-64-0
MDL Number: MFCD00077479
Molecular Formula: C7H3ClF2O2
Molecular Weight: 192.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 105

Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
Boiling Pt: 258
Melting Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00077479
CAS Number: 110877-64-0
EINECS: 405-380-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e6723fdcbec9a37bec7c8b6c65ee0681554fa5f&vid=de5d099e87898c2bb7ddc3297555756cab124d13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR