tert-Butyl cyanoacetate ≥97.0%
Supplier: TCI America
C1441-250ML
C1441-25ML
TCC1441-250MLEA
598.82
CAD
TCC1441-250ML
TCC1441-025ML
tert-Butyl cyanoacetate ≥97.0%
tert-Butyl cyanoacetate
CAS Number: 1116-98-9
MDL Number: MFCD00001938
Molecular Formula: C7H11NO2
Molecular Weight: 141.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 91
Specific Gravity (20/20): 0.99
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Cyanoacetic Acid tert-Butyl Ester
Formula:
C₇H₁₁NO₂ Boiling Pt: 108 °C Density: 0.99 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD00001938 CAS Number: 1116-98-9 EINECS: 214-243-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...