2-Chloronicotinamide ≥98.0%

Supplier: TCI America
Warning

C1404-5G C1404-25G
TCC1404-5GEA 66.36 CAD
TCC1404-5G TCC1404-025G
2-Chloronicotinamide ≥98.0%
2-Chloronicotinamide

CAS Number: 10366-35-5
MDL Number: MFCD00006237
Molecular Formula: C6H5ClN2O
Molecular Weight: 156.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 164


Other Product Names / Synonyms: 2-Chloropyridine-3-carboxamide

Formula: C₆H₅ClN₂O
Melting Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00006237
CAS Number: 10366-35-5
EINECS: 233-808-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=461eb750bc5f84b4b625c3647cc3634c7b5ee397&vid=461eb750bc5f84b4b625c3647cc3634c7b5ee397&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR