2-Chlorobenzoxazole ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloro-1,3-benzoxazole

C1387-5G C1387-25G
TCC1387-005GEA 48.24 CAD
TCC1387-005G TCC1387-025G
2-Chlorobenzoxazole ≥98.0%
2-Chlorobenzoxazole

CAS Number: 615-18-9
MDL Number: MFCD00005766
Molecular Formula: C7H4ClNO
Molecular Weight: 153.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 202
Melting point (°C): 7
Flash Point (°C): 85
Specific Gravity (20/20): 1.32

Formula: C₇H₄ClNO
MW: 153.57 g/mol
Boiling Pt: 202 °C
Melting Pt: 7 °C
Density: 1.32 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00005766
CAS Number: 615-18-9
EINECS: 210-414-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=516cbc3dec9fa9f4e24af8c1548d0af52c508e7a&vid=516cbc3dec9fa9f4e24af8c1548d0af52c508e7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR