1,1'-Thiocarbonyldiimidazole ≥95.0% (by titrimetric analysis)

Supplier: TCI America

C1376-25G C1376-5G
TCC1376-25GEA 810.86 CAD
TCC1376-25G TCC1376-005G
1,1'-Thiocarbonyldiimidazole ≥95.0% (by titrimetric analysis)
1,1'-Thiocarbonyldiimidazole

CAS Number: 6160-65-2
MDL Number: MFCD00005289
Molecular Formula: C7H6N4S
Molecular Weight: 178.21
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 102
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Phosgene & Thiophosgene Alternatives
Other Product Names / Synonyms: 1,1'-Carbonothioylbis-1H-imidazole

Formula: C₇H₆N₄S
Melting Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005289
CAS Number: 6160-65-2
EINECS: 228-183-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa0c85d130a0fdc76f646a2c8292728ce34fa8dd&vid=fa0c85d130a0fdc76f646a2c8292728ce34fa8dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR