Cyclohexyl phenyl ketone ≥98.0%

Supplier: TCI America

C1345-25G C1345-500G
TCC1345-025GEA 88.43 CAD
TCC1345-025G TCC1345-500G
Cyclohexyl phenyl ketone ≥98.0%
Cyclohexyl phenyl ketone

CAS Number: 712-50-5
MDL Number: MFCD00001467
Molecular Formula: C13H16O
Molecular Weight: 188.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 168
Melting point (°C): 56
Flash Point (°C): 110


Other Product Names / Synonyms: Benzoylcyclohexane

Formula: C₁₃H₁₆O
Boiling Pt: 168 °C
Melting Pt: 56 °C
Flash Pt: 110 °C
MDL Number: MFCD00001467
CAS Number: 712-50-5
EINECS: 211-923-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb5a9f14cccc2808362975367ae8df554f902bae&vid=bb5a9f14cccc2808362975367ae8df554f902bae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR