ω,4-Dichloroacetophenone ≥98.0%

Supplier: TCI America

Synonyms: α,4-Dichloroacetophenone, 2,4'-Dichloroacetophenone

C1306-5G C1306-25G
TCC1306-005GEA 115.34 CAD
TCC1306-005G TCC1306-025G
ω,4-Dichloroacetophenone ≥98.0%
ω,4-Dichloroacetophenone

CAS Number: 937-20-2
MDL Number: MFCD00018926
Molecular Formula: C8H6Cl2O
Molecular Weight: 189.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 270
Melting point (°C): 101


Other Product Names / Synonyms: 2,4'-Dichloroacetophenone

Formula: C₈H₆Cl₂O
Boiling Pt: 270 °C
Melting Pt: 101 °C
MDL Number: MFCD00018926
CAS Number: 937-20-2
EINECS: 213-323-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fed5a926990f1036daec42e385856dd12a484b06&vid=fed5a926990f1036daec42e385856dd12a484b06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR