4-(4-Chlorophenyl)piperidin-4-ol 97,0

Supplier: TCI America

Synonyms: 4-(4-Chlorophenyl)-4-hydroxypiperidine

C1291-25G C1291-5G
TCC1291-025GEA 303.36 CAD
TCC1291-025G TCC1291-005G
4-(4-Chlorophenyl)piperidin-4-ol 97,0
4-(4-Chlorophenyl)piperidin-4-ol

CAS Number: 39512-49-7
MDL Number: MFCD00006001
Molecular Formula: C11H14ClNO
Molecular Weight: 211.69
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 141


Other Product Names / Synonyms: 4-(4-Chlorophenyl)-4-piperidinol

Formula: C₁₁H₁₄ClNO
MW: 211.69 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00006001
CAS Number: 39512-49-7
EINECS: 254-479-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee0285a9c74faa58eafc380511146276399875c5&vid=ee0285a9c74faa58eafc380511146276399875c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR