3-Hydroxypropiononitrile ≥95.0%

Supplier: TCI America

C1285-500ML C1285-25ML
TCC1285-500MLEA 388.68 CAD
TCC1285-500ML TCC1285-025ML
3-Hydroxypropiononitrile ≥95.0%
3-Hydroxypropiononitrile

CAS Number: 109-78-4
MDL Number: MFCD00002826
Molecular Formula: C3H5NO
Molecular Weight: 71.08
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 221
Flash Point (°C): 137
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 2-Cyanoethanol, Hydracrylonitrile, 3-Hydroxypropionitrile

Formula: C₃H₅NO
Boiling Pt: 221 °C
Melting Pt: –46 °C
Density: 1.05 (20 °C)
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00002826
CAS Number: 109-78-4
EINECS: 203-704-8
Merck Index: 13,03828
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c994066873bf6895548669ce9edfe61f16f51399&vid=c994066873bf6895548669ce9edfe61f16f51399&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR