3-Chloro-4-fluorobenzoic acid ≥98.0%
Supplier: TCI America
C1275-5G
C1275-25G
TCC1275-005GEA
58.46
CAD
TCC1275-005G
TCC1275-25G
3-Chloro-4-fluorobenzoic acid ≥98.0%
3-Chloro-4-fluorobenzoic acid
CAS Number: 403-16-7
MDL Number: MFCD00042477
Molecular Formula: C7H4ClFO2
Molecular Weight: 174.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 134
Formula:
C₇H₄ClFO₂ MW: 174.56 g/mol Boiling Pt: 273 Melting Pt: 136 |
MDL Number:
MFCD00042477 CAS Number: 403-16-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...