(2-Cyanoethyl)triethoxysilane ≥98.0%

Supplier: TCI America

C1259-250ML C1259-25ML
TCC1259-250MLEA 553 CAD
TCC1259-250ML TCC1259-025ML
(2-Cyanoethyl)triethoxysilane ≥98.0%
(2-Cyanoethyl)triethoxysilane

CAS Number: 919-31-3
MDL Number: MFCD00019851
Molecular Formula: C9H19NO3Si
Molecular Weight: 217.34
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 86
Specific Gravity (20/20): 0.98


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: 3-(Triethoxysilyl)propionitrile

Formula: NCCH₂CH₂Si(OCH₂CH₃)₃
Boiling Pt: 137 °C
Density: 0.98 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00019851
CAS Number: 919-31-3
EINECS: 213-050-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a413f2d248aeaed6771d021f47938fae0c57cde9&vid=a413f2d248aeaed6771d021f47938fae0c57cde9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR